This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00903637_m1
See other IFNLR1 GE Assays ›
Gene Symbol:
IFNLR1
Gene Name:
interferon lambda receptor 1
Gene Aliases:
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2
Chromosome Location:
Chr.1: 24154156 - 24187275 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00903637_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFNLR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170743.3 NP_734464.1 2 - 3 205 126
NM_173064.2 NP_775087.1 2 - 3 205 126
NM_173065.2 NP_775088.1 2 - 3 205 126
GenBank mRNA AF439325.1 - 2 - 3 178 126
AJ534330.1 - 2 - 3 191 126
AJ534331.1 - 2 - 3 191 126
AK160364.1 - 2 - 3 181 126
AY129151.1 - 2 - 3 178 126
AY129152.1 - 2 - 3 178 126
AY129153.1 - 2 - 3 178 126
BC101975.1 - 2 - 3 191 126
BC139722.1 - 2 - 3 191 126
BC140872.1 - 2 - 3 254 9021740 126
CN289861.1 - 2 - 3 205 126
JN591376.1 - 2 - 3 178 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100013315.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002359.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFNLR1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

mucosal immune response
negative regulation of cell proliferation
cytokine-mediated signaling pathway
response to type III interferon
regulation of defense response to virus by host
defense response to virus
cytokine receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 ENST00000327535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 ENST00000327575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 ENST00000374421.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036464.hg.1 TC01002359.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198888.hg.1 TC0100013315.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0