Assay Details
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244889.1 | NP_001231818.1 | 9 - 10 | 2103 | 114 | |
NM_170744.4 | NP_734465.2 | 10 - 11 | 2136 | 114 | ||
XM_011539453.1 | XP_011537755.1 | 11 - 12 | 1685 | 114 | ||
XM_017015834.1 | XP_016871323.1 | 11 - 12 | 1695 | 114 | ||
XM_017015835.1 | XP_016871324.1 | 10 - 11 | 2488 | 114 | ||
GenBank mRNA | AB096256.1 | - | 10 - 11 | 2091 | 114 | |
AK022859.1 | - | 1 - 2 | 151 | 114 | ||
AK094595.1 | - | 3 - 4 | 606 | 114 | ||
AK128132.1 | - | 10 - 11 | 2136 | 114 | ||
AY126437.1 | - | 9 - 10 | 1711 | 114 | ||
AY358351.1 | - | 10 - 11 | 2082 | 114 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000438.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK022859.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
apoptotic process
signal transduction
positive regulation of phosphatidylinositol 3-kinase signaling
anterior/posterior axon guidance
negative regulation of neuron apoptotic process
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding
angiogenesis
apoptotic process
signal transduction
positive regulation of phosphatidylinositol 3-kinase signaling
anterior/posterior axon guidance
negative regulation of neuron apoptotic process
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10005135.hg.1 | ENST00000335350.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10005135.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10005135.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10005135.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10005135.hg.1 | TC10000438.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007950.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | ENST00000335350.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | ENST00000373192.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | NM_001244889.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | NM_170744.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000153956.hg.1 | TC1000007950.hg.1 | Clariom D TC |
