This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00898202_g1
See other TM9SF4 GE Assays ›
Gene Symbol:
TM9SF4
Gene Name:
transmembrane 9 superfamily member 4
Gene Aliases:
dJ836N17.2
Chromosome Location:
Chr.20: 32109506 - 32167258 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00898202_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TM9SF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014742.3 NP_055557.2 7 - 8 1010 64
XM_005260622.4 XP_005260679.1 7 - 8 1062 64
XM_017028154.1 XP_016883643.1 7 - 8 791 64
XM_017028155.1 XP_016883644.1 7 - 8 1062 64
XM_017028156.1 XP_016883645.1 7 - 8 758 64
XM_017028157.1 XP_016883646.1 7 - 8 1015 64
XM_017028158.1 XP_016883647.1 7 - 8 753 64
XM_017028159.1 XP_016883648.1 7 - 8 815 64
XM_017028160.1 XP_016883649.1 7 - 8 727 64
XM_017028161.1 XP_016883650.1 7 - 8 763 64
GenBank mRNA AK126534.1 - 5 - 6 460 64
AK294980.1 - 7 - 8 758 64
AK296500.1 - 7 - 8 667 64
AK302439.1 - 8 - 9 865 64
AK316293.1 - 6 - 7 714 64
BC021107.1 - 7 - 8 786 4896167 64
BC022850.2 - 6 - 7 768 64
D87444.1 - 7 - 8 1064 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007105.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000207.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017028159.1,XM_017028160.1,XM_017028161.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TM9SF4

Panther Classification:

Molecular Function -

transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20002542.hg.1 ENST00000217315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20002542.hg.1 ENST00000398022.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20002542.hg.1 ENST00000421148.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20002542.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20002542.hg.1 TC20000207.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay PSR2000148557.hg.1 TC2000007105.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0