This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00897488_s1
See other EGFLAM-AS2 GE Assays ›
Gene Symbol:
EGFLAM-AS2
Gene Name:
EGFLAM antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 38399814 - 38403471 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00897488_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EGFLAM-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102749.1 2 601 98
GenBank mRNA AK093278.1 - 2 601 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500010534.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173404.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173404.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173404.hg.1 TC0500010534.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173406.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173406.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173406.hg.1 TC0500010534.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500173405.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173405.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173405.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173405.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173405.hg.1 TC0500010534.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 229269 - -
Array Name Probe IDs Targets Target Type
Illumina human-6 v2.0 expression beadchip ILMN_1827152 - -
Illumina Sentrix Human-6 v2 Expression BeadChip 5310671 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_97789 - -
Illumina HumanWG-6_V2_0_R2 ILMN_1827152 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0