This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00895549_g1
See other COL9A2 GE Assays ›
Gene Symbol:
COL9A2
Gene Name:
collagen type IX alpha 2
Gene Aliases:
DJ39G22.4, EDM2, MED, STL5
Chromosome Location:
Chr.1: 40300487 - 40317653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00895549_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COL9A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001852.3 NP_001843.1 10 - 11 592 90
XM_006710365.3 XP_006710428.1 11 - 12 700 90
XM_011540717.2 XP_011539019.1 9 - 10 1508 90
XM_017000333.1 XP_016855822.1 9 - 10 646 90
GenBank mRNA BC041479.1 - 9 - 10 825 90
BC136326.1 - 10 - 11 585 9020313 90
BC136327.1 - 10 - 11 585 9020314 90
M95610.1 - 10 - 11 481 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100013823.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002554.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011540717.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COL9A2

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
extracellular matrix organization
extracellular matrix structural constituent conferring tensile strength

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372736.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372736.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372736.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372736.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372736.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000372748.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000417105.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000427563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000461118.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000466267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000482722.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000488463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000490132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000490132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000490132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000490132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000490132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000495948.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 ENST00000496215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013823.hg.1 NM_001852.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0