This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00893125_m1
See other SOS1 GE Assays ›
Gene Symbol:
SOS1
Gene Name:
SOS Ras/Rac guanine nucleotide exchange factor 1
Gene Aliases:
GF1, GGF1, GINGF, HGF, NS4
Chromosome Location:
Chr.2: 38981549 - 39124959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00893125_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005633.3 NP_005624.2 11 - 12 1982 94
XM_005264515.4 XP_005264572.1 11 - 12 2211 94
XM_011533062.2 XP_011531364.1 11 - 12 2150 94
XM_011533064.2 XP_011531366.1 11 - 12 1890 94
XM_011533066.2 XP_011531368.1 4 - 5 891 94
GenBank mRNA AB209140.1 - 4 - 5 953 94
AK290228.1 - 12 - 13 2028 94
L13857.1 - 11 - 12 1941 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012307.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001761.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SOS1

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
B cell homeostasis
hair follicle development
cardiac atrium morphogenesis
pericardium morphogenesis
signal transduction
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
vitellogenesis
axon guidance
insulin receptor signaling pathway
regulation of T cell differentiation in thymus
regulation of Rho protein signal transduction
multicellular organism growth
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
regulation of T cell proliferation
positive regulation of apoptotic process
positive regulation of GTPase activity
positive regulation of epidermal growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
blood vessel morphogenesis
leukocyte migration
regulation of small GTPase mediated signal transduction
positive regulation of small GTPase mediated signal transduction
palate development
eyelid development in camera-type eye
heart trabecula morphogenesis
midbrain morphogenesis
regulation of pro-B cell differentiation
DNA binding
guanyl-nucleotide exchange factor activity
Ras guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
SH3 domain binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000395038.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000402219.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000426016.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000451331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000451331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000451331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000451331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000461545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000461545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000461545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000461545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000469581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000469581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000469581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000472480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000472480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000474390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000474390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000474390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 ENST00000474390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012307.hg.1 NM_005633.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0