This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00864332_m1
See other LOC101929073 GE Assays ›
Gene Symbol:
LOC101929073
Gene Name:
uncharacterized LOC101929073
Gene Aliases:
-
Chromosome Location:
Chr.10: 25924378 - 25933710 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00864332_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929073
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL137590.1 - 1 - 2 163 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 836 LINC00836 -191.4kb Anti-sense Chr. 10 - 25651712 - 25732935 on NCBI Build GRCh38
Downstream myosin IIIA MYO3A DFNB30 5.8kb Anti-sense Chr. 10 - 25934073 - 26212536 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000552511.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000552511.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008828.hg.1 ENST00000552511.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000548631.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550118.2 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000550680.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000552511.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000552511.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008828.hg.1 ENST00000552511.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0