This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00851991_u1
See other RPL5 GE Assays ›
Gene Symbol:
RPL5
Gene Name:
ribosomal protein L5
Gene Aliases:
DBA6, L5, MSTP030, PPP1R135
Chromosome Location:
Chr.1: 92832037 - 92841924 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00851991_u1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000969.3 NP_000960.2 2 - 3 152 134
GenBank mRNA AF113210.1 - 2 - 3 131 134
AK222604.1 - 2 - 3 116 134
AK314720.1 - 2 - 3 149 134
BC001882.1 - 2 - 3 118 134
BC109370.1 - 2 - 3 136 5736022 134
BC132970.1 - 2 - 3 121 40146992 134
BC132972.1 - 2 - 3 121 40146994 134
BG756697.1 - 2 - 3 137 134
BU941919.1 - 2 - 3 153 134
U14966.1 - 2 - 3 104 133

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF113210.1, AK314720.1, BC001882.1, BC109370.1, BC132970.1, BC132972.1, NM_000969.3, U14966.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL5

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
RNA binding
structural constituent of ribosome
protein binding
5S rRNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007599.hg.1 ENST00000509786.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007599.hg.1 ENST00000509786.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007588.hg.1 ENST00000600819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007588.hg.1 ENST00000600819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014969.hg.1 ENST00000444235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008975.hg.1 ENST00000399168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012382.hg.1 ENST00000418634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012382.hg.1 ENST00000418634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007482.hg.1 ENST00000567049.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008208.hg.1 ENST00000477071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007058.hg.1 ENST00000494197.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010589.hg.1 ENST00000504890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009010.hg.1 ENST00000437011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009010.hg.1 ENST00000437011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000383953.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000384792.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NR_000006.8 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NR_002444.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015640.hg.1 ENST00000396820.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009010.hg.1 ENST00000437011.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009010.hg.1 ENST00000437011.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012382.hg.1 ENST00000418634.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012382.hg.1 ENST00000418634.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010589.hg.1 ENST00000504890.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007482.hg.1 ENST00000567049.3 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000383953.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000384792.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NR_000006.8 Refseq
Affymetrix Human Clariom D Assay TC0100009046.hg.1 NR_002444.2 Refseq
Affymetrix Human Clariom D Assay TC0500007599.hg.1 ENST00000509786.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007599.hg.1 ENST00000509786.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008975.hg.1 ENST00000399168.2 Ensembl
Affymetrix Human Clariom D Assay TC0200015640.hg.1 ENST00000396820.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007588.hg.1 ENST00000600819.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007588.hg.1 ENST00000600819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008208.hg.1 ENST00000477071.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007058.hg.1 ENST00000494197.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Clariom D Assay TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Clariom D Assay TC2200009120.hg.1 ENST00000398869.2 Ensembl
Affymetrix Human Clariom D Assay TC0200014969.hg.1 ENST00000444235.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 115662 - -
ABI Human Genome Survey Microarray Version 2.0 164436 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0