Assay Details
Gene Symbol: NDUFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004542.3 | NP_004533.1 | 2 | 49 | 147 | |
XM_017026833.1 | XP_016882322.1 | 3 | 466 | 147 | ||
GenBank mRNA | AF044955.1 | - | 2 | 41 | 147 | |
AF070653.1 | - | 2 | 50 | 147 | ||
AK289489.1 | - | 2 | 47 | 147 | ||
BC022369.1 | - | 2 | 46 | 4737656 | 147 | |
BC061644.1 | - | 2 | 49 | 5748578 | 147 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000871.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF044955.1,AF070653.1,AK289489.1,BC022369.1,BC061644.1,NM_004542.3,XM_017026833.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600007520.hg.1 | ENST00000571486.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011660.hg.1 | ENST00000391762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011660.hg.1 | ENST00000485876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011660.hg.1 | NM_004542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011660.hg.1 | TC19000871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011775.hg.1 | ENST00000423475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | ENST00000391762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | ENST00000391764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | ENST00000417328.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | ENST00000485876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | NM_004542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011657.hg.1 | TC19000871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011661.hg.1 | ENST00000391762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011661.hg.1 | ENST00000417328.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011661.hg.1 | ENST00000485876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011661.hg.1 | NM_004542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011661.hg.1 | TC19000871.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100011775.hg.1 | ENST00000423475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007520.hg.1 | ENST00000571486.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900202542.hg.1 | TC1900011795.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000303553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391763.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000391764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000417903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000420296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000451517.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000471292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000484103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011795.hg.1 | ENST00000485876.5 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 189650 | - | - |
