This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00543910_m1
See other IL1RL2 GE Assays ›
Gene Symbol:
IL1RL2
Gene Name:
interleukin 1 receptor like 2
Gene Aliases:
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2
Chromosome Location:
Chr.2: 102186973 - 102242906 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00543910_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1RL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003854.2 NP_003845.2 2 - 3 190 76
XM_005264035.1 XP_005264092.1 2 - 3 183 76
XM_005264036.1 XP_005264093.1 2 - 3 105 76
XM_011512091.1 XP_011510393.1 2 - 3 183 76
XM_011512092.1 XP_011510394.1 2 - 3 183 76
XM_011512093.1 XP_011510395.1 2 - 3 183 76
XM_011512094.1 XP_011510396.1 2 - 3 105 76
XM_011512096.1 XP_011510398.1 2 - 3 183 76
XM_017005174.1 XP_016860663.1 2 - 3 183 76
XM_017005177.1 XP_016860666.1 2 - 3 266 76
XM_017005178.1 XP_016860667.1 2 - 3 132 76
XM_017005180.1 XP_016860669.1 1 - 2 68 76
GenBank mRNA AF284434.1 - 2 - 3 198 76
AK315334.1 - 2 - 3 105 76
U49065.1 - 2 - 3 198 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008669.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000621.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017005177.1,XM_017005178.1,XM_017005180.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL1RL2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cellular defense response
signal transduction
positive regulation of interleukin-6 production
innate immune response
positive regulation of T cell differentiation
regulation of inflammatory response
interleukin-1-mediated signaling pathway
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02009859.hg.1 ENST00000264257.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009859.hg.1 ENST00000539491.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009859.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009859.hg.1 TC02000621.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160542.hg.1 TC0200008669.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0