This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00542560_m1
See other CAMK2G GE Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00542560_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001320898.1
NP_001307827.1
18 - 19
1428
77
NM_172171.2
NP_751911.1
17 - 18
1401
77
XM_005270195.1
XP_005270252.1
18 - 19
1455
77
XM_005270197.1
XP_005270254.1
18 - 19
1419
77
XM_005270198.1
XP_005270255.1
18 - 19
1413
77
XM_005270199.1
XP_005270256.1
17 - 18
1386
77
XM_005270200.1
XP_005270257.1
17 - 18
1356
77
XM_005270201.1
XP_005270258.1
17 - 18
1350
77
XM_006717993.1
XP_006718056.1
19 - 20
1488
77
XM_006717996.1
XP_006718059.1
18 - 19
1425
77
XM_011540225.2
XP_011538527.1
17 - 18
1340
77
XM_017016716.1
XP_016872205.1
19 - 20
1488
77
XM_017016717.1
XP_016872206.1
19 - 20
1436
77
XM_017016718.1
XP_016872207.1
18 - 19
1455
77
XM_017016719.1
XP_016872208.1
19 - 20
1446
77
XM_017016720.1
XP_016872209.1
18 - 19
1425
77
XM_017016721.1
XP_016872210.1
19 - 20
1563
77
XM_017016722.1
XP_016872211.1
18 - 19
1413
77
XM_017016723.1
XP_016872212.1
17 - 18
1392
77
XM_017016724.1
XP_016872213.1
17 - 18
1386
77
XM_017016725.1
XP_016872214.1
17 - 18
1356
77
XM_017016726.1
XP_016872215.1
17 - 18
1350
77
XM_017016727.1
XP_016872216.1
16 - 17
1323
77
XM_017016728.1
XP_016872217.1
17 - 18
1340
77
XM_017016729.1
XP_016872218.1
17 - 18
1223
77
XM_017016731.1
XP_016872220.1
17 - 18
1331
77
XM_017016732.1
XP_016872221.1
17 - 18
1261
77
XM_017016734.1
XP_016872223.1
17 - 18
1351
77
XM_017016737.1
XP_016872226.1
16 - 17
1294
77
XM_017016738.1
XP_016872227.1
18 - 19
1354
77
XM_017016739.1
XP_016872228.1
19 - 20
1620
77
GenBank mRNA
AK131199.1
-
17 - 18
1324
77
U50359.1
-
6 - 7
299
77
U66063.1
-
7 - 8
446
77
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Protein Kinase
Protein Phosphatase
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene CAMK2G
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0