This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00411819_m1
See other SNX29 GE Assays ›
Gene Symbol:
SNX29
Gene Name:
sorting nexin 29
Gene Aliases:
A-388D4.1, RUNDC2A
Chromosome Location:
Chr.16: 11976734 - 12574289 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00411819_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNX29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032167.4 NP_115543.3 17 - 18 2029 82
XM_005255682.3 XP_005255739.1 17 - 18 2054 82
XM_011522738.2 XP_011521040.1 17 - 18 2201 82
XM_011522741.2 XP_011521043.1 17 - 18 2185 82
XM_011522743.2 XP_011521045.1 17 - 18 2189 82
XM_011522744.2 XP_011521046.1 17 - 18 2060 82
XM_017023871.1 XP_016879360.1 17 - 18 2198 82
XM_017023872.1 XP_016879361.1 17 - 18 2018 82
XM_017023873.1 XP_016879362.1 17 - 18 2185 82
XM_017023874.1 XP_016879363.1 17 - 18 2018 82
XM_017023875.1 XP_016879364.1 15 - 16 1725 82
XM_017023878.1 XP_016879367.1 7 - 8 1523 82
GenBank mRNA AK074072.1 - 7 - 8 636 82
BC029857.1 - 5 - 6 423 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006941.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000167.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SNX29

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Process(es)

phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0