This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00392254_gH
See other DIP2C GE Assays ›
Gene Symbol:
DIP2C
Gene Name:
disco interacting protein 2 homolog C
Gene Aliases:
KIAA0934
Chromosome Location:
Chr.10: 274190 - 689710 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00392254_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIP2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014974.2 NP_055789.1 21 - 22 2680 75
XM_005252426.3 XP_005252483.1 22 - 23 2839 75
XM_005252427.3 XP_005252484.1 21 - 22 2720 75
XM_005252428.4 XP_005252485.1 22 - 23 2890 75
XM_005252430.2 XP_005252487.2 5 - 6 930 75
XM_011519428.2 XP_011517730.1 22 - 23 2821 75
XM_011519429.2 XP_011517731.1 22 - 23 2819 75
XM_011519430.2 XP_011517732.1 21 - 22 2696 75
XM_011519431.2 XP_011517733.1 21 - 22 2696 75
XM_011519432.1 XP_011517734.1 21 - 22 3121 75
XM_011519433.2 XP_011517735.1 5 - 6 1929 75
XM_017015977.1 XP_016871466.1 5 - 6 941 75
GenBank mRNA AB023151.2 - 21 - 22 2678 75
AK126063.1 - 23 - 24 2933 75
AK298353.1 - 5 - 6 777 75
AK309715.1 - 5 - 6 692 75
AK310018.1 - 4 - 5 933 75
BC063313.1 - 21 - 22 2680 30349267 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009516.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000961.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DIP2C

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
molecular_function
catalytic activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000280886.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000381496.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000421992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000423550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 ENST00000434695.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009516.hg.1 NM_014974.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0