This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00388731_g1
See other TRADD GE Assays ›
Gene Symbol:
TRADD
Gene Name:
TNFRSF1A associated via death domain
Gene Aliases:
Hs.89862
Chromosome Location:
Chr.16: 67154185 - 67159909 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00388731_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRADD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323552.1 NP_001310481.1 3 - 4 524 67
NM_003789.3 NP_003780.1 3 - 4 511 67
XM_005256213.3 XP_005256270.1 3 - 4 1028 67
XM_017023815.1 XP_016879304.1 3 - 4 577 67
GenBank mRNA AK090673.1 - 1 - 2 875 67
AK315654.1 - 3 - 4 904 67
BC004491.2 - 3 - 4 488 3689007 67
BG717460.1 - 3 - 4 506 67
BT006934.1 - 2 - 3 431 67
DA353575.1 - 3 - 4 511 67
L41690.1 - 3 - 4 479 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600010616.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001180.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRADD

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
I-kappaB kinase/NF-kappaB signaling
extrinsic apoptotic signaling pathway via death domain receptors
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
tumor necrosis factor-mediated signaling pathway
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
positive regulation of hair follicle development
death-inducing signaling complex assembly
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
signal transducer activity
tumor necrosis factor receptor binding
protein binding
kinase binding
protein complex binding
identical protein binding
binding, bridging
death domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16016390.hg.1 ENST00000345057.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016390.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016390.hg.1 TC16001180.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016389.hg.1 ENST00000345057.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16016389.hg.1 NM_003789.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16016389.hg.1 TC16001180.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay PSR1600181662.hg.1 TC1600010616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000345057.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000486556.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 ENST00000566247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 NM_003789.3 Refseq
Affymetrix Human Clariom D Assay PSR1600181660.hg.1 TC1600010616.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0