This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00386020_m1
See other NECAB2 GE Assays ›
Gene Symbol:
NECAB2
Gene Name:
N-terminal EF-hand calcium binding protein 2
Gene Aliases:
EFCBP2
Chromosome Location:
Chr.16: 83968632 - 84002776 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00386020_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NECAB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019065.2 NP_061938.2 10 - 11 980 121
XM_011523135.1 XP_011521437.1 9 - 10 797 121
XM_017023317.1 XP_016878806.1 8 - 9 1835 121
GenBank mRNA AF070637.1 - 6 - 7 575 121
AF193758.1 - 6 - 7 573 121
AK127376.1 - 10 - 11 901 121
AY299331.1 - 10 - 11 980 121
BC016979.1 - 8 - 9 976 121
BC131615.1 - 5 - 6 466 40116488 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008633.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000653.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NECAB2

Gene Ontology Categories:

Process(es)

calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16009202.hg.1 ENST00000305202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009202.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009202.hg.1 TC16000653.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16009203.hg.1 ENST00000305202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009203.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009203.hg.1 TC16000653.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay TC1600008633.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164458.hg.1 TC1600008633.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000566836.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 ENST00000567703.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164455.hg.1 TC1600008633.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000305202.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000564166.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 ENST00000565691.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 NM_019065.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164459.hg.1 TC1600008633.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0