This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00382327_g1
See other RBBP8NL GE Assays ›
Gene Symbol:
RBBP8NL
Gene Name:
RBBP8 N-terminal like
Gene Aliases:
C20orf151
Chromosome Location:
Chr.20: 62410237 - 62427573 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00382327_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBBP8NL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080833.2 NP_543023.2 10 - 11 1729 100
GenBank mRNA AK074919.1 - 10 - 11 1689 100
AK128718.1 - 1 - 2 1109 100
AK290299.1 - 10 - 11 1702 100
BC006450.1 - 3 - 4 934 100
BC033142.1 - 3 - 4 934 100
BC137326.1 - 9 - 10 1617 9021324 100
BC137327.1 - 9 - 10 1617 9021325 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009751.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20001010.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBBP8NL

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20013870.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20013870.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20013870.hg.1 TC20001010.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay PSR2000168018.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168018.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay PSR2000168018.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168018.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay PSR2000168018.hg.1 TC2000009751.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 ENST00000252998.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq
Affymetrix Human Clariom D Assay TC2000009751.hg.1 NM_080833.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0