This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00381179_g1
See other SLC25A16 GE Assays ›
Gene Symbol:
SLC25A16
Gene Name:
solute carrier family 25 member 16
Gene Aliases:
D10S105E, GDA, GDC, HGT.1, ML7, hML7
Chromosome Location:
Chr.10: 68479961 - 68527523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00381179_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324312.1 NP_001311241.1 5 - 6 689 112
NM_001324313.1 NP_001311242.1 5 - 6 689 112
NM_001324314.1 NP_001311243.1 6 - 7 871 112
NM_001324315.1 NP_001311244.1 5 - 6 1546 112
NM_001324317.1 NP_001311246.1 4 - 5 555 112
NM_152707.3 NP_689920.1 5 - 6 689 112
XM_011540211.2 XP_011538513.1 5 - 6 689 112
GenBank mRNA AK290255.1 - 5 - 6 618 112
AK295287.1 - 5 - 6 654 112
AK298510.1 - 6 - 7 764 112
BC030266.1 - 5 - 6 640 5241247 112
M31659.1 - 6 - 7 708 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010849.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001349.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC25A16

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

translation
transport
coenzyme biosynthetic process
transmembrane transport
structural constituent of ribosome
secondary active transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000439904.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000439904.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0