This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00375497_g1
See other MRPL24 GE Assays ›
Gene Symbol:
MRPL24
Gene Name:
mitochondrial ribosomal protein L24
Gene Aliases:
L24mt, MRP-L18, MRP-L24
Chromosome Location:
Chr.1: 156737302 - 156741267 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00375497_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024540.3 NP_078816.2 2 - 3 323 96
NM_145729.2 NP_663781.1 2 - 3 364 96
XM_011509981.2 XP_011508283.1 2 - 3 434 96
XM_011509982.2 XP_011508284.1 2 - 3 332 96
GenBank mRNA AK024570.1 - 2 - 3 321 96
AK222500.1 - 2 - 3 321 96
AK223066.1 - 2 - 3 321 96
BC012440.1 - 2 - 3 305 3863359 96
BC016700.1 - 2 - 3 323 4108596 96
BG706700.1 - 2 - 3 328 96
CR457329.1 - 1 - 2 184 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016021.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003352.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRPL24

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
mitochondrial translational elongation
mitochondrial translational termination
structural constituent of ribosome

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01052347.hg.1 ENST00000361531.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01052347.hg.1 ENST00000368211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01052347.hg.1 NM_024540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01052347.hg.1 NM_145729.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01052347.hg.1 TC01003352.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay PSR0100221569.hg.1 TC0100016021.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000361531.6 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000368211.8 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000412846.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000420938.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000434558.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 ENST00000478899.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_024540.3 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq
Affymetrix Human Clariom D Assay TC0100016021.hg.1 NM_145729.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0