This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00343016_s1
See other LOC93622 GE Assays ›
Gene Symbol:
LOC93622
Gene Name:
Morf4 family associated protein 1 like 1 pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.4: 6674094 - 6676047 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00343016_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC93622
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015433.1 94
GenBank mRNA AK026375.1 - 94
AK314832.1 - 94
BC006130.2 - 94
BC021542.1 - 94
BC026970.1 - 94
BC040997.1 - 94
BC066911.1 - 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK314832.1 transcripts.

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0