This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00298360_m1
See other EPB41L4B GE Assays ›
Gene Symbol:
EPB41L4B
Gene Name:
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases:
CG1, EHM2
Chromosome Location:
Chr.9: 109171974 - 109320964 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00298360_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41L4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019114.4 NP_061987.3 24 - 25 3003 137
XM_011518790.1 XP_011517092.1 25 - 26 2623 137
XM_011518791.2 XP_011517093.1 25 - 26 2599 137
XM_017014813.1 XP_016870302.1 24 - 25 2494 137
GenBank mRNA AB209187.1 - 16 - 17 1595 137
AF153416.1 - 24 - 25 2775 137
AL353947.1 - 4 - 5 454 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011160.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPB41L4B

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
regulation of keratinocyte proliferation
actomyosin structure organization
wound healing
positive regulation of cell adhesion
positive regulation of keratinocyte migration
structural constituent of cytoskeleton
cytoskeletal protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay PSR0900175904.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175904.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay PSR0900175904.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175904.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay PSR0900175904.hg.1 TC0900011160.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay TC0900011160.hg.1 NM_019114.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 170260 - -
ABI Human Genome Survey Microarray Version 2.0 185297 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0