This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00273714_s1
See other RPA4 GE Assays ›
Gene Symbol:
RPA4
Gene Name:
replication protein A4
Gene Aliases:
HSU24186
Chromosome Location:
Chr.X: 96883908 - 96885467 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00273714_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013347.4 NP_037479.1 1 194 74
GenBank mRNA BC069791.1 - 1 89 7262610 74
BC069824.1 - 1 89 7262622 74
BC104013.2 - 1 65 40024430 74
BC104014.2 - 1 65 40024431 74
U24186.2 - 1 194 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007890.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000465.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC069791.1,BC069824.1,BC104013.2,BC104014.2,NM_013347.4,U24186.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPA4

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
G1/S transition of mitotic cell cycle
DNA replication initiation
nucleotide-excision repair
DNA recombination
nuclear DNA replication
single-stranded DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X006007.hg.1 ENST00000373040.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X006007.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X006007.hg.1 TC0X000465.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X006005.hg.1 ENST00000373040.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X006005.hg.1 NM_013347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X006005.hg.1 TC0X000465.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00153667.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153667.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153667.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153667.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153667.hg.1 TC0X00007890.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay TC0X00007890.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153670.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153670.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153670.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153670.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153670.hg.1 TC0X00007890.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00153669.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153669.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153669.hg.1 ENST00000373040.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153669.hg.1 NM_013347.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00153669.hg.1 TC0X00007890.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 205623 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0