This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00170197_m1
See other HTRA1 GE Assays ›
Gene Symbol:
HTRA1
Gene Name:
HtrA serine peptidase 1
Gene Aliases:
ARMD7, CADASIL2, CARASIL, HtrA, L56, ORF480, PRSS11
Chromosome Location:
Chr.10: 122461525 - 122514908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00170197_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HTRA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002775.4 NP_002766.1 1 - 2 602 95
GenBank mRNA AF097709.1 - 1 - 2 45 95
AK091944.1 - 3 - 4 377 95
AK092476.1 - 2 - 3 383 95
BC011352.1 - 2 - 3 369 95
BC031082.1 - 1 - 2 510 95
D87258.1 - 1 - 2 522 95
DA633502.1 - 3 - 4 377 95
Y07921.1 - 1 - 2 512 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009152.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000889.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK092476.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens 1700 Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HTRA1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
placenta development
proteolysis
extracellular matrix disassembly
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
positive regulation of epithelial cell proliferation
negative regulation of defense response to virus
chorionic trophoblast cell differentiation
dentinogenesis
serine-type endopeptidase activity
insulin-like growth factor binding
serine-type peptidase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10010825.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010825.hg.1 ENST00000435263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010825.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010825.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010829.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010829.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010829.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010830.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010830.hg.1 ENST00000435263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010830.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010830.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010827.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010827.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010827.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010828.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010828.hg.1 ENST00000435263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010828.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010828.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10010831.hg.1 ENST00000368984.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010831.hg.1 ENST00000435263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010831.hg.1 NM_002775.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010831.hg.1 TC10000889.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000162710.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162710.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162710.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162710.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162710.hg.1 TC1000009152.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000162708.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162708.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162708.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162708.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162708.hg.1 TC1000009152.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 ENST00000420892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay TC1000009152.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162711.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162711.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162711.hg.1 ENST00000368984.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000162711.hg.1 NM_002775.4 Refseq
Affymetrix Human Clariom D Assay PSR1000162711.hg.1 TC1000009152.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 111748 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0