This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05258537_m1
See other JHY GE Assays ›
Gene Symbol:
Jhy
Gene Name:
junctional cadherin complex regulator
Gene Aliases:
CUNH11orf63
Chromosome Location:
Chr.Un|NW_004624880: 2150509 - 2218204 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05258537_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Jhy
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004870901.3 XP_004870958.1 5 - 6 1420 69
XM_004870902.3 XP_004870959.1 5 - 6 1674 69
XM_004870903.3 XP_004870960.1 4 - 5 1133 69
XM_004870904.3 XP_004870961.1 4 - 5 1382 69
XM_013067859.2 XP_012923313.1 5 - 6 1674 69
XM_013067860.2 XP_012923314.1 5 - 6 1674 69
XM_021249041.1 XP_021104700.1 5 - 6 1674 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0