This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05243687_m1
See other LOC101713038 GE Assays ›
Gene Symbol:
LOC101713038
Gene Name:
src kinase-associated phosphoprotein 2
Gene Aliases:
Skap2
Chromosome Location:
Chr.Un|NW_004624739: 3668630 - 3850571 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05243687_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101713038
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013070279.2 XP_012925733.1 8 - 9 785 90
XM_013070280.2 XP_012925734.1 8 - 9 785 90
XM_021254096.1 XP_021109755.1 8 - 9 755 90
XM_021254097.1 XP_021109756.1 8 - 9 888 90
XM_021254098.1 XP_021109757.1 8 - 9 848 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0