This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05208792_g1
See other DNLZ GE Assays ›
Gene Symbol:
Dnlz
Gene Name:
DNL-type zinc finger
Gene Aliases:
-
Chromosome Location:
-
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05208792_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnlz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013073871.2 XP_012929325.1 1 - 2 316 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0