This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05186131_m1
See other LOC101723006 GE Assays ›
Gene Symbol:
LOC101723006
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
Luc7l2
Chromosome Location:
Chr.Un|NW_004624765: 19768008 - 19851985 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05186131_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101723006
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004851264.3 XP_004851321.1 2 - 3 329 87
XM_004851266.3 XP_004851323.1 3 - 4 1587 87
XM_004851267.3 XP_004851324.1 3 - 4 225 87
XM_004851269.3 XP_004851326.1 2 - 3 468 87
XM_021263677.1 XP_021119336.1 4 - 5 312 87
XM_021263678.1 XP_021119337.1 4 - 5 315 87
XM_021263679.1 XP_021119338.1 5 - 6 388 87
XM_021263680.1 XP_021119339.1 3 - 4 227 87
XM_021263681.1 XP_021119340.1 4 - 5 319 87
XM_021263682.1 XP_021119341.1 3 - 4 2630 87
XM_021263683.1 XP_021119342.1 3 - 4 1281 87
XM_021263684.1 XP_021119343.1 3 - 4 1393 87
XM_021263685.1 XP_021119344.1 2 - 3 112 87
XM_021263686.1 XP_021119345.1 4 - 5 354 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021263677.1, XM_021263678.1, XM_021263682.1, XM_021263683.1, XM_021263684.1, XM_021263685.1, XM_021263686.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0