Assay Details
Gene Symbol: C2cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021237513.1 | XP_021093172.1 | 11 - 12 | 1208 | 173 | |
XM_021237514.1 | XP_021093173.1 | 10 - 11 | 1163 | 173 | ||
XM_021237515.1 | XP_021093174.1 | 12 - 13 | 1184 | 173 | ||
XM_021237517.1 | XP_021093176.1 | 11 - 12 | 1208 | 173 |

More Information
Set Membership: |
![]() ![]() |
