This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05141659_mH
See other SLC9A3R2 GE Assays ›
Gene Symbol:
Slc9a3r2
Gene Name:
SLC9A3 regulator 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624913: 210325 - 220805 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05141659_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9a3r2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004873664.3 XP_004873721.1 4 - 5 893 60
XM_004873666.3 XP_004873723.1 3 - 4 608 60
XM_004873669.3 XP_004873726.1 4 - 5 895 60
XM_013068921.2 XP_012924375.1 4 - 5 896 60
XM_021251359.1 XP_021107018.1 4 - 5 898 60
XM_021251360.1 XP_021107019.1 3 - 4 655 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0