Assay Details
Gene Symbol: Mbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867548.3 | XP_004867605.1 | 7 - 8 | 965 | 77 | |
XM_013066635.2 | XP_012922089.1 | 7 - 8 | 1003 | 77 | ||
XM_013066636.2 | XP_012922090.1 | 8 - 9 | 1078 | 77 | ||
XM_021246583.1 | XP_021102242.1 | 7 - 8 | 1004 | 77 | ||
XM_021246584.1 | XP_021102243.1 | 7 - 8 | 988 | 77 | ||
XM_021246585.1 | XP_021102244.1 | 7 - 8 | 967 | 77 | ||
XM_021246586.1 | XP_021102245.1 | 8 - 9 | 1045 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
