This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05116621_g1
See other ACTR3B GE Assays ›
Gene Symbol:
Actr3b
Gene Name:
ARP3 actin related protein 3 homolog B
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624800: 4257792 - 4322692 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05116621_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actr3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004860348.2 XP_004860405.1 11 - 12 1255 71
XM_004860349.2 XP_004860406.1 9 - 10 1051 71
XM_004860350.3 XP_004860407.1 10 - 11 1233 71
XM_021240802.1 XP_021096461.1 11 - 12 1135 71
XM_021240804.1 XP_021096463.1 11 - 12 1195 71
XM_021240805.1 XP_021096464.1 9 - 10 931 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0