This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05107541_m1
See other ELP1 GE Assays ›
Gene Symbol:
Elp1
Gene Name:
elongator complex protein 1
Gene Aliases:
GW7_20449, Ikbkap
Chromosome Location:
Chr.Un|NW_004624758: 2575313 - 2646313 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05107541_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004848154.2 XP_004848211.1 5 - 6 658 58
XM_004848159.3 XP_004848216.1 2 - 3 156 58
XM_021261149.1 XP_021116808.1 6 - 7 821 58
XM_021261150.1 XP_021116809.1 6 - 7 764 58
XM_021261151.1 XP_021116810.1 2 - 3 308 58
XM_021261152.1 XP_021116811.1 2 - 3 281 58
XM_021261153.1 XP_021116812.1 2 - 3 100 58
XM_021261154.1 XP_021116813.1 2 - 3 96 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0