Assay Details
Gene Symbol: Lyrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847750.3 | XP_004847807.1 | 1 - 2 | 580 | 88 | |
XM_004847751.2 | XP_004847808.1 | 1 - 2 | 564 | 88 | ||
XM_013073268.2 | XP_012928722.1 | 1 - 2 | 579 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
