This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07186760_s1
See other GPRC5B GE Assays ›
Gene Symbol:
GPRC5B
Gene Name:
G protein-coupled receptor class C group 5 member B
Gene Aliases:
-
Chromosome Location:
Chr.14: 15029188 - 15046207 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07186760_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004945464.3 XP_004945521.1 3 5218 71
XM_015294793.2 XP_015150279.1 4 4872 71
XM_015294794.1 XP_015150280.1 4 4513 71
XM_015294795.1 XP_015150281.1 5 4628 71
XM_015294797.2 XP_015150283.1 4 4884 71
XM_015294798.1 XP_015150284.1 4 4509 71
GenBank mRNA GU724343.1 - 1 1218 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0