This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07181743_s1
See other RGL4 GE Assays ›
Gene Symbol:
RGL4 GTF3C5
Gene Name:
ral guanine nucleotide dissociation stimulator like 4
general transcription factor IIIC subunit 5
Gene Aliases:
RALGDS
-
Chromosome Location:
Chr.17: 7088198 - 7127900 on Build GRCg6a
Chromosome Location:
Chr.17: 7080377 - 7089081 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
-
Assay ID Gg07181743_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015279767.2 XP_015135253.2 18 4303 102
XM_015279768.2 XP_015135254.2 18 4300 102
XM_015279769.2 XP_015135255.2 18 4270 102
XM_015279770.2 XP_015135256.2 18 4189 102
XM_015279771.2 XP_015135257.2 18 4039 102
XM_015279772.2 XP_015135258.2 18 3859 102
XM_015279773.2 XP_015135259.2 18 3956 102
XM_015279774.2 XP_015135260.2 18 3815 102
XM_015279775.2 XP_015135261.2 18 4186 102
XM_025141410.1 XP_024997178.1 18 4036 102
XM_025141411.1 XP_024997179.1 18 3810 102
Gene Symbol: GTF3C5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_025156063.1 XP_025011831.1 11 3557 102
XM_025156064.1 XP_025011832.1 10 3414 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor RNA metabolism protein nucleic acid metabolism protein general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0