This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07181726_s1
See other B2M GE Assays ›
Gene Symbol:
B2M
Gene Name:
beta-2-microglobulin
Gene Aliases:
b2m-W01, b2m-W03, b2m-Z01, b2m-Z02, b2m-Z03
Chromosome Location:
Chr.10: 1784065 - 1786517 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07181726_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B2M
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001750.3 NP_001001750.2 4 1587 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of T cell mediated cytotoxicity
response to molecule of bacterial origin
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
positive regulation of T cell cytokine production
iron ion transport
immune response
learning or memory
negative regulation of neuron projection development
antigen processing and presentation of endogenous peptide antigen via MHC class I
T cell differentiation in thymus
regulation of iron ion transport
protein refolding
regulation of erythrocyte differentiation
positive regulation of receptor-mediated endocytosis
negative regulation of epithelial cell proliferation
protein homotetramerization
iron ion homeostasis
cellular response to iron(III) ion
cellular response to nicotine
modulation of age-related behavioral decline
negative regulation of receptor binding
positive regulation of ferrous iron binding
positive regulation of transferrin receptor binding
amyloid fibril formation
positive regulation of cellular senescence
negative regulation of forebrain neuron differentiation
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0