This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07179126_m1
See other TFDP3 GE Assays ›
Gene Symbol:
TFDP3
Gene Name:
transcription factor Dp family member 3
Gene Aliases:
TFDP1
Chromosome Location:
Chr.1: 138648961 - 138690364 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07179126_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TFDP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004938530.3 XP_004938587.1 7 - 8 772 135
XM_015277902.2 XP_015133388.1 8 - 9 912 135
XM_015277903.2 XP_015133389.1 8 - 9 914 135
XM_015277904.2 XP_015133390.1 8 - 9 909 135
XM_015277905.2 XP_015133391.1 8 - 9 911 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0