This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07156639_m1
See other MRPS26 GE Assays ›
Gene Symbol:
MRPS26
Gene Name:
mitochondrial ribosomal protein S26
Gene Aliases:
GNRH2, GnRHII
Chromosome Location:
Chr.4: 88643965 - 88649469 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07156639_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPS26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039913.1 NP_001035002.1 4 - 5 783 97
XM_015286027.2 XP_015141513.2 4 - 5 940 97
GenBank mRNA AB194405.1 - 4 - 5 783 97
AB194406.1 - 2 - 3 388 97
AB194407.1 - 1 - 2 233 97
BX932366.1 - 2 - 3 391 97
BX934126.1 - 2 - 3 357 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

peptide hormone ribosomal protein intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0