This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03815778_s1
See other TPM2 GE Assays ›
Gene Symbol:
TPM2
Gene Name:
tropomyosin 2
Gene Aliases:
BRT-2, TPM3, beta-tropomyosin, tropomyosin-2
Chromosome Location:
Chr.Z: 9207199 - 9218220 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03815778_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205446.1 NP_990777.1 8 1052 108
XM_015277261.2 XP_015132747.1 9 1212 108
XM_015277263.1 XP_015132749.1 9 1206 108
XM_015277269.2 XP_015132755.1 8 1231 108
GenBank mRNA M23081.1 - 1437 108
M64287.1 - 8 1052 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

muscle contraction
actin filament organization
protein binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header