This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03815143_s1
See other PITX2 GE Assays ›
Gene Symbol:
PITX2
Gene Name:
paired like homeodomain 2
Gene Aliases:
Ptx2
Chromosome Location:
Chr.4: 57785032 - 57798939 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03815143_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205010.1 NP_990341.1 3 1550 85
XM_025149515.1 XP_025005283.1 4 1370 85
XM_025149516.1 XP_025005284.1 4 1384 85
GenBank mRNA AF076640.1 - 3 1550 85
BX935055.1 - 4 1160 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

heart looping
heart morphogenesis
determination of left/right symmetry
anatomical structure morphogenesis
embryonic limb morphogenesis
response to retinoic acid
positive regulation of myoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic organ morphogenesis
positive regulation of dermatome development
positive regulation of myotome development
positive regulation of myoblast proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0