This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03814244_s1
See other CDKN1A GE Assays ›
Gene Symbol:
CDKN1A
Gene Name:
cyclin dependent kinase inhibitor 1A
Gene Aliases:
CIP1
Chromosome Location:
Chr.26: 1600491 - 1602395 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03814244_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204396.1 NP_989727.1 1248 82
GenBank mRNA AF513031.1 - 1248 82
BX936101.2 - 599 82
CR385180.1 - 992 82
CR406048.1 - 622 82
CR406257.1 - 575 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase inhibitor kinase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0