Assay Details
Gene Symbol: TTC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006552.1 | NP_001006552.1 | 10 - 11 | 1315 | 69 | |
GenBank mRNA | AJ720886.1 | - | 10 - 11 | 1315 | 69 | |
CR354321.1 | - | 8 - 9 | 959 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
