This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03367098_m1
See other CSRP2 GE Assays ›
Gene Symbol:
CSRP2
Gene Name:
cysteine and glycine rich protein 2
Gene Aliases:
BETA-CRP, CRP2
Chromosome Location:
Chr.1: 38319984 - 38327780 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03367098_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSRP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205208.1 NP_990539.1 2 - 3 139 69
XM_015279087.2 XP_015134573.1 2 - 3 214 69
XM_025146679.1 XP_025002447.1 2 - 3 273 69
GenBank mRNA FJ882406.1 - 2 - 3 146 69
X84264.1 - 139 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
actin cytoskeleton organization
sarcomere organization
muscle tissue development
structural constituent of muscle
actinin binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0