This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03366417_m1
See other TFAP2A GE Assays ›
Gene Symbol:
TFAP2A
Gene Name:
transcription factor AP-2 alpha
Gene Aliases:
TFAP2
Chromosome Location:
Chr.2: 63283222 - 63300911 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03366417_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TFAP2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205094.1 NP_990425.1 3 - 4 625 61
XM_015275844.2 XP_015131330.1 3 - 4 2013 61
XM_015275845.2 XP_015131331.1 3 - 4 2368 61
XM_015275846.2 XP_015131332.1 3 - 4 1142 61
XM_015275847.2 XP_015131333.1 4 - 5 810 61
XM_015275848.2 XP_015131334.1 3 - 4 839 61
GenBank mRNA U72992.1 - 3 - 4 625 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header