This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03366210_m1
See other MTFR1 GE Assays ›
Gene Symbol:
MTFR1
Gene Name:
mitochondrial fission regulator 1
Gene Aliases:
CHPPR
Chromosome Location:
Chr.2: 114804131 - 114825447 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03366210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204543.2 NP_989874.2 6 - 7 987 108
XM_015282720.2 XP_015138206.1 6 - 7 1017 108
XM_015282721.2 XP_015138207.1 6 - 7 1022 108
XM_015282722.2 XP_015138208.1 6 - 7 1012 108
XM_015282723.2 XP_015138209.1 6 - 7 885 108
XM_015282724.2 XP_015138210.1 6 - 7 938 108
XM_025147021.1 XP_025002789.1 6 - 7 985 108
GenBank mRNA AF208489.1 - 6 - 7 923 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s)

mitochondrial fission
mitochondrion organization
aerobic respiration

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0