This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363760_m1
See other CHRNA7 GE Assays ›
Gene Symbol:
CHRNA7
Gene Name:
cholinergic receptor nicotinic alpha 7 subunit
Gene Aliases:
-
Chromosome Location:
Chr.10: 7109509 - 7149541 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363760_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHRNA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204181.2 NP_989512.2 5 - 6 542 62
GenBank mRNA X52295.1 - 436 62
X68586.1 - 540 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
response to hypoxia
calcium ion transport
cellular calcium ion homeostasis
signal transduction
synaptic transmission, cholinergic
positive regulation of cell proliferation
negative regulation of tumor necrosis factor production
ion transmembrane transport
response to nicotine
positive regulation of angiogenesis
cognition
excitatory postsynaptic potential
beta-amyloid binding
acetylcholine receptor activity
toxic substance binding
chloride channel regulator activity
acetylcholine-gated cation channel activity
acetylcholine binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0