This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363363_m1
See other RUNX2 GE Assays ›
Gene Symbol:
RUNX2
Gene Name:
runt related transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 109294565 - 109509337 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363363_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RUNX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204128.1 NP_989459.1 5 - 6 770 97
XM_015285080.2 XP_015140566.1 6 - 7 1933 97
XM_015285081.2 XP_015140567.1 4 - 5 742 97
XM_025148466.1 XP_025004234.1 5 - 6 2152 97
GenBank mRNA AF445419.1 - 5 - 6 770 97
AF450085.1 - 5 - 6 770 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

Runt transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0