This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03362470_m1
See other ALDH1A1 GE Assays ›
Gene Symbol:
ALDH1A1
Gene Name:
aldehyde dehydrogenase 1 family member A1
Gene Aliases:
GGADHR
Chromosome Location:
Chr.Z: 35905454 - 35958939 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03362470_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204577.4 NP_989908.1 10 - 11 2740 62
XM_015280233.2 XP_015135719.1 7 - 8 879 62
GenBank mRNA X58869.4 - 2740 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
retinol metabolic process
oxidation-reduction process
retinal dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
benzaldehyde dehydrogenase (NAD+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0