This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03362334_m1
See other HDAC1 GE Assays ›
Gene Symbol:
HDAC1
Gene Name:
histone deacetylase 1
Gene Aliases:
-
Chromosome Location:
Chr.23: 5272426 - 5285757 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03362334_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204156.1 NP_989487.1 4 - 5 431 71
GenBank mRNA AF039751.1 - 4 - 5 416 71
AF043328.1 - 4 - 5 406 71
AF044169.1 - 4 - 5 406 71
AJ453995.1 - 4 - 5 431 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin organization
methylation-dependent chromatin silencing
endoderm development
positive regulation of cell proliferation
epidermal cell differentiation
positive regulation of receptor biosynthetic process
neuron differentiation
circadian regulation of gene expression
embryonic digit morphogenesis
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation by host of viral transcription
positive regulation of transcription from RNA polymerase II promoter
positive regulation of oligodendrocyte differentiation
regulation of endopeptidase activity
negative regulation of androgen receptor signaling pathway
eyelid development in camera-type eye
fungiform papilla formation
histone H3 deacetylation
histone H4 deacetylation
negative regulation of canonical Wnt signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
RNA polymerase II repressing transcription factor binding
p53 binding
transcription corepressor activity
histone deacetylase activity
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
activating transcription factor binding
Krueppel-associated box domain binding
histone deacetylase binding
protein N-terminus binding
NF-kappaB binding
E-box binding
promoter-specific chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0