This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03360184_s1
See other CCR5 GE Assays ›
Gene Symbol:
CCR5
Gene Name:
C-C motif chemokine receptor 5
Gene Aliases:
CCR1-L
Chromosome Location:
Chr.2: 42616854 - 42639981 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03360184_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271141.1 NP_001258070.1 3 1255 115
XM_015281502.2 XP_015136988.1 5 1543 115
XM_025147218.1 XP_025002986.1 3 1308 115
XM_025147219.1 XP_025002987.1 3 1233 115
XM_025147220.1 XP_025002988.1 4 1377 115
GenBank mRNA EF524205.1 - 2 1200 115
FR874035.1 - 1 953 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0