This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03358992_m1
See other GNAL GE Assays ›
Gene Symbol:
GNAL
Gene Name:
G protein subunit alpha L
Gene Aliases:
-
Chromosome Location:
Chr.2: 97333366 - 97516571 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03358992_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008746.1 NP_001008746.1 11 - 12 1154 105
XM_015282481.2 XP_015137967.1 11 - 12 1551 105
XM_015282482.2 XP_015137968.1 12 - 13 1328 105
XM_025147343.1 XP_025003111.1 11 - 12 1107 105
GenBank mRNA AY091502.1 - 11 - 12 1041 105
BX931962.1 - 11 - 12 1154 105
CR353558.1 - 12 - 13 946 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0